missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,3,4,6-Tetra-O-benzyl-D-glucopyranose, 98+%

Catalog No. p-7030319
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2788803 1 g
AAH2788806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2788803 Supplier Thermo Scientific Chemicals Supplier No. H2788803
Only null left

CAS: 4132-28-9 | C34H36O6 | 540.656 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in chloroform

Notes
Store -20°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 4132-28-9
Molecular Formula C34H36O6
Molecular Weight (g/mol) 540.656
MDL Number MFCD00066004
InChI Key OGOMAWHSXRDAKZ-BKJHVTENSA-N
Synonym 2,3,4,6-tetra-o-benzyl-d-glucopyranose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl tetrahydro-2h-pyran-2-ol, tetrabenzylglucose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl oxan-2-ol, d-glucopyranose, 2,3,4,6-tetrakis-o-phenylmethyl, ksc235o0p, 2,3,4,6-tetra-o-benzylglucopyranose, 2,3,4,6-tetra-o-benzyl glucopyranose, 2,3,4,6-tetra-o-benzyl-glucopyranose, 2,3,4,6-terta-o-benzyl-d-glucopyranose
PubChem CID 10940502
IUPAC Name (3R,4S,5R,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5

Specifications

Melting Point 150°C to 155°C
Quantity 1 g
Solubility Information Soluble in chloroform
Formula Weight 540.65
Percent Purity ≥98%
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-D-glucopyranose
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only