missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-benzoyl-beta-D-glucopyranosyl Isothiocyanate 98.0+%, TCI America™

Catalog No. A55151G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55151G 1 g
A5515100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55151G Supplier TCI America Supplier No. A55151G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132413-50-4
Molecular Formula C35H27NO9S
Molecular Weight (g/mol) 637.66
MDL Number MFCD00075064
InChI Key RITGYQZMPNVVEG-UHFFFAOYNA-N
Synonym BGIT
PubChem CID 11966661
IUPAC Name [3,4,5-tris(benzoyloxy)-6-isothiocyanatooxan-2-yl]methyl benzoate
SMILES O=C(OCC1OC(N=C=S)C(OC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 637.66
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-benzoyl-beta-D-glucopyranosyl Isothiocyanate [for HPLC Labeling]