accessibility menu, dialog, popup

UserName

OOONCSOOOOOO

CAS RN 132413-50-4

IUPAC Name: [3,4,5-tris(benzoyloxy)-6-isothiocyanatooxan-2-yl]methyl benzoate
Synonyms: [3,4,5-tris(benzoyloxy)-6-isothiocyanatooxan-2-yl]methyl benzoate
Molecular Weight (g/mol): 637.66
Molecular Formula: C35H27NO9S
InChi Key: RITGYQZMPNVVEG-UHFFFAOYNA-N
SMILES: O=C(OCC1OC(N=C=S)C(OC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results