missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-acetyl-D-mannopyranosyl Fluoride 95.0+%, TCI America™

Supplier:  TCI America T25675G

 View more versions of this product

Catalog No. T25675G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3,4,6-Tetra-O-acetyl-D-mannopyranosyl Fluoride
White-Yellow
5 g
CC(=O)OCC1C(C(C(C(O1)F)OC(=O)C)OC(=O)C)OC(=O)C
350.295
350.30
Crystalline Powder
174511-17-2
C14H19FO9
JJXATNWYELAACC-JABUTEAWSA-N
[(2R,3R,4S,5S)-3,4,5-triacetyloxy-6-fluorooxan-2-yl]methyl acetate
45489774
≥95.0% (GC)

Chemical Identifiers

174511-17-2
350.295
45489774
CC(=O)OCC1C(C(C(C(O1)F)OC(=O)C)OC(=O)C)OC(=O)C
C14H19FO9
JJXATNWYELAACC-JABUTEAWSA-N
[(2R,3R,4S,5S)-3,4,5-triacetyloxy-6-fluorooxan-2-yl]methyl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions