missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-acetyl-D-mannopyranosyl Fluoride 95.0+%, TCI America™

Catalog No. p-7137287
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25671G 1 g
T25675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25671G Supplier TCI America Supplier No. T25671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 174511-17-2
Molecular Formula C14H19FO9
Molecular Weight (g/mol) 350.295
InChI Key JJXATNWYELAACC-JABUTEAWSA-N
PubChem CID 45489774
IUPAC Name [(2R,3R,4S,5S)-3,4,5-triacetyloxy-6-fluorooxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)F)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 350.30
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-D-mannopyranosyl Fluoride
TSCA No
Recommended Storage Refrigerator