Learn More
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate is a chiral derivatization reagent which reacts mainly with enantiomeric amino acids.
Supplier:  Merck Emd Millipore T57831G
 View more versions of this product
Catalog No. 111013073
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
114°C to 116°C
Bottomless Glass Bottle with Inserted Fused Cone
MFCD00043085
GITC
≥98.0% (HPLC)
14152-97-7
C15H19 NO9 S
1 g
389.38
For chiral derivatization
Chemical Identifiers
MFCD00043085
GITC
Product Suggestions
Product Suggestions