missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187674
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013074 100 mg
111013073 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013074 Supplier Merck Emd Millipore Supplier No. T5783100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate is a chiral derivatization reagent which reacts mainly with enantiomeric amino acids.

Chemical Identifiers

MDL Number MFCD00043085
Synonym GITC

Specifications

Melting Point 114°C to 116°C
CAS 14152-97-7
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C15H19 NO9 S
Quantity 100 mg
Formula Weight 389.38
Percent Purity ≥98.0% (HPLC)
Grade For chiral derivatization