missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetraphenylthiophene 98.0+%, TCI America™

Supplier:  TCI America T220025G

 View more versions of this product

Catalog No. T220025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3,4,5-Tetraphenylthiophene
186°C
C28H20S
25 g
MQFBWJOMLIHUDY-UHFFFAOYSA-N
2,3,4,5-tetraphenylthiophene
74664
≥98.0% (GC)
1884-68-0
Yellow
MFCD00014527
tetraphenylthiophene, thiophene, tetraphenyl, thionessal, acmc-1bvi3, 2,4,5-tetraphenylthiophene, thiophene,2,3,4,5-tetraphenyl
C1=CC=C(C=C1)C2=C(SC(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5
388.528
388.53
Crystalline Powder

Chemical Identifiers

1884-68-0
388.528
MQFBWJOMLIHUDY-UHFFFAOYSA-N
74664
C1=CC=C(C=C1)C2=C(SC(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5
C28H20S
MFCD00014527
tetraphenylthiophene, thiophene, tetraphenyl, thionessal, acmc-1bvi3, 2,4,5-tetraphenylthiophene, thiophene,2,3,4,5-tetraphenyl
2,3,4,5-tetraphenylthiophene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions