missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4,5-Tetraphenylthiophene 98.0+%, TCI America™
$76.43 - $374.68
Chemical Identifiers
| CAS | 1884-68-0 |
|---|---|
| Molecular Formula | C28H20S |
| Molecular Weight (g/mol) | 388.528 |
| MDL Number | MFCD00014527 |
| InChI Key | MQFBWJOMLIHUDY-UHFFFAOYSA-N |
| Synonym | tetraphenylthiophene, thiophene, tetraphenyl, thionessal, acmc-1bvi3, 2,4,5-tetraphenylthiophene, thiophene,2,3,4,5-tetraphenyl |
| PubChem CID | 74664 |
| IUPAC Name | 2,3,4,5-tetraphenylthiophene |
| SMILES | C1=CC=C(C=C1)C2=C(SC(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 |
Chemical Identifiers
| 1884-68-0 | |
| 388.528 | |
| MQFBWJOMLIHUDY-UHFFFAOYSA-N | |
| 74664 | |
| C1=CC=C(C=C1)C2=C(SC(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 |
| C28H20S | |
| MFCD00014527 | |
| tetraphenylthiophene, thiophene, tetraphenyl, thionessal, acmc-1bvi3, 2,4,5-tetraphenylthiophene, thiophene,2,3,4,5-tetraphenyl | |
| 2,3,4,5-tetraphenylthiophene |
Specifications
| 1884-68-0 | |
| Yellow | |
| MFCD00014527 | |
| tetraphenylthiophene, thiophene, tetraphenyl, thionessal, acmc-1bvi3, 2,4,5-tetraphenylthiophene, thiophene,2,3,4,5-tetraphenyl | |
| C1=CC=C(C=C1)C2=C(SC(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 | |
| 388.528 | |
| 388.53 | |
| Crystalline Powder |
| 186°C | |
| C28H20S | |
| 5 g | |
| MQFBWJOMLIHUDY-UHFFFAOYSA-N | |
| 2,3,4,5-tetraphenylthiophene | |
| 74664 | |
| ≥98.0% (GC) | |
| 2,3,4,5-Tetraphenylthiophene |
Safety and Handling
TSCA : No