missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4,5-Tetrafluorobenzoyl Chloride 97.0+%, TCI America™
$30.80 - $84.70
Chemical Identifiers
| CAS | 94695-48-4 |
|---|---|
| Molecular Formula | C7HClF4O |
| Molecular Weight (g/mol) | 212.53 |
| MDL Number | MFCD00075164 |
| InChI Key | XWCKIXLTBNGIHV-UHFFFAOYSA-N |
| Synonym | 2,3,4,5-tetrafluorobenzoylchloride, tetrafluorobenzoyl chloride, 2,3,4,5-tetrafluoro benzoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro, 2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobezoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, 2,3,4,5-tetrafluorobezoylchloride |
| PubChem CID | 2733689 |
| IUPAC Name | 2,3,4,5-tetrafluorobenzoyl chloride |
| SMILES | FC1=CC(C(Cl)=O)=C(F)C(F)=C1F |
Chemical Identifiers
| 94695-48-4 | |
| 212.53 | |
| XWCKIXLTBNGIHV-UHFFFAOYSA-N | |
| 2733689 | |
| FC1=CC(C(Cl)=O)=C(F)C(F)=C1F |
| C7HClF4O | |
| MFCD00075164 | |
| 2,3,4,5-tetrafluorobenzoylchloride, tetrafluorobenzoyl chloride, 2,3,4,5-tetrafluoro benzoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro, 2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobezoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, 2,3,4,5-tetrafluorobezoylchloride | |
| 2,3,4,5-tetrafluorobenzoyl chloride |
Specifications
| 94695-48-4 | |
| C7HClF4O | |
| 5 g | |
| 2,3,4,5-tetrafluorobenzoylchloride, tetrafluorobenzoyl chloride, 2,3,4,5-tetrafluoro benzoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro, 2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobezoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, 2,3,4,5-tetrafluorobezoylchloride | |
| FC1=CC(C(Cl)=O)=C(F)C(F)=C1F | |
| 212.53 | |
| 212.53 | |
| Liquid |
| Yellow | |
| MFCD00075164 | |
| 3265 | |
| XWCKIXLTBNGIHV-UHFFFAOYSA-N | |
| 2,3,4,5-tetrafluorobenzoyl chloride | |
| 2733689 | |
| ≥97.0% (GC,T) | |
| 2,3,4,5-Tetrafluorobenzoyl Chloride |
Safety and Handling
TSCA : No