missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4,5-Tetrafluorobenzaldehyde 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 82°C |
| Quantity | 1 g |
| UN Number | 1989 |
| Formula Weight | 178.09 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,3,4,5-Tetrafluorobenzaldehyde |
Safety and Handling
| TSCA | No |