missing translation for 'onlineSavingsMsg'
Learn More

2',3,4,5-Tetrafluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™

Catalog No. p-7137692
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33171G 1 g
T33175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33171G Supplier TCI America Supplier No. T33171G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 173837-35-9
Molecular Formula C21H22F4
Molecular Weight (g/mol) 350.401
MDL Number MFCD22380673
InChI Key AYFPNRLYKGMWJN-UHFFFAOYSA-N
PubChem CID 12010666
IUPAC Name 1,2,3-trifluoro-5-[2-fluoro-4-(4-propylcyclohexyl)phenyl]benzene
SMILES CCCC1CCC(CC1)C2=CC(=C(C=C2)C3=CC(=C(C(=C3)F)F)F)F

Specifications

Melting Point 63°C
Color White
Quantity 1 g
Formula Weight 350.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3,4,5-Tetrafluoro-4′-(trans-4-propylcyclohexyl)biphenyl