accessibility menu, dialog, popup

UserName

CAS RN 173837-35-9

IUPAC Name: 2,3',4',5'-tetrafluoro-4-(4-propylcyclohexyl)-1,1'-biphenyl
Synonyms: 2,3',4',5'-tetrafluoro-4-(4-propylcyclohexyl)-1,1'-biphenyl
Molecular Weight (g/mol): 350.4
Molecular Formula: C21H22F4
InChi Key: AYFPNRLYKGMWJN-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1=CC(F)=C(C=C1)C1=CC(F)=C(F)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results