missing translation for 'onlineSavingsMsg'
Learn More

2',3,4,5-Tetrafluoro-4″-propyl-1,1':4',1″-terphenyl 98.0+%, TCI America™

Catalog No. T33181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33181G 1 g
T33185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33181G Supplier TCI America Supplier No. T33181G

Chemical Identifiers

CAS 205806-87-7
Molecular Formula C21H16F4
Molecular Weight (g/mol) 344.35
MDL Number MFCD12964272
InChI Key YBPXWWAJMUUUAV-UHFFFAOYSA-N
PubChem CID 12010664
IUPAC Name 2,3',4',5'-tetrafluoro-4-(4-propylphenyl)-1,1'-biphenyl
SMILES CCCC1=CC=C(C=C1)C1=CC(F)=C(C=C1)C1=CC(F)=C(F)C(F)=C1

Specifications

Color White
Quantity 1 g
Synonym 2′C,3,4,5-Tetrafluoro-4′C′C-propyl-p-terphenyl
Formula Weight 344.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3,4,5-Tetrafluoro-4′′-propyl-1,1′:4′,1′′-terphenyl