missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrachloronitrobenzene 98.0+%, TCI America™

Catalog No. T11281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11281G Supplier TCI America Supplier No. T11281G
Only null left

Chemical Identifiers

CAS 879-39-0
Molecular Formula C6HCl4NO2
Molecular Weight (g/mol) 260.88
MDL Number MFCD00007064
InChI Key MTBYTWZDRVOMBR-UHFFFAOYSA-N
PubChem CID 13428
IUPAC Name 1,2,3,4-tetrachloro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Cl)=C(Cl)C(Cl)=C1Cl

Specifications

Melting Point 65°C
Color Yellow
Quantity 1 g
Formula Weight 260.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,5-Tetrachloronitrobenzene
RTECSNumber DB9625000
TSCA No