missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,5-Di-O-isopropylidene-D-arabitol, 98%, Thermo Scientific Chemicals

Catalog No. AAJ6660803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6660803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6660803 Supplier Thermo Scientific Chemicals Supplier No. J6660803
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Protect from heat. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 19139-74-3
Molecular Formula C11H20O5
Molecular Weight (g/mol) 232.28
MDL Number MFCD00063387
InChI Key FBIWGPWXTDOFQR-UHFFFAOYNA-N
Synonym 2-o,3-o:4-o,5-o-diisopropylidene-d-arabinitol
PubChem CID 11042611
IUPAC Name [(4R,5R)-5-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(C)OCC(O1)C1OC(C)(C)OC1CO

Specifications

Color Colorless
Quantity 1 g
Formula Weight 232.27
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 2,3:4,5-Di-O-isopropylidene-D-arabitol
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only