accessibility menu, dialog, popup

UserName

H3COOOOH3CH3COHCH3

CAS RN 19139-74-3

IUPAC Name: [5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
Synonyms: [5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
Molecular Weight (g/mol): 232.28
Molecular Formula: C11H20O5
InChi Key: FBIWGPWXTDOFQR-UHFFFAOYNA-N
SMILES: CC1(C)OCC(O1)C1OC(C)(C)OC1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results