missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorostyrene (stabilized with TBC) 98.0+%, TCI America™

Catalog No. p-7134817
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P08625G 5 g
P086225G 25 g
2 options

Catalog No. P08625G

Supplier: TCI America P08625G

Only null left

Chemical Identifiers

CAS 653-34-9
Molecular Formula C8H3F5
Molecular Weight (g/mol) 194.104
MDL Number MFCD00000300
InChI Key LVJZCPNIJXVIAT-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorostyrene, pentafluorostyrene, benzene, ethenylpentafluoro, 1,2,3,4,5-pentafluoro-6-vinyl-benzene, 2',3',4',5',6'-pentafluorostyrene, 2-2,3,4,5,6-pentafluorophenyl ethyl, vinylpentafluorobenzene, acmc-1avmt, styrene, 2,3,4,5,6-pentafluoro, 1,2,3,4,5-pentafluoro-6-vinylbenzene
PubChem CID 69556
IUPAC Name 1-ethenyl-2,3,4,5,6-pentafluorobenzene
SMILES C=CC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
UN Number 1993
Formula Weight 194.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4,5,6-Pentafluorostyrene (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator