Learn More
2,3,4,5,6-Pentafluorophenylacetic acid, 98+%
CAS: 653-21-4 | C8H3F5O2 | 226.102 g/mol
$78.56 - $250.67
Chemical Identifiers
| CAS | 653-21-4 |
|---|---|
| Molecular Formula | C8H3F5O2 |
| Molecular Weight (g/mol) | 226.102 |
| MDL Number | MFCD00004316 |
| InChI Key | LGCODSNZJOVMHV-UHFFFAOYSA-N |
| Synonym | pentafluorophenylacetic acid, 2,3,4,5,6-pentafluorophenylacetic acid, pentafluorophenyl acetic acid, 2-perfluorophenyl acetic acid, 2-2,3,4,5,6-pentafluorophenyl acetic acid, 2,3,4,5,6-pentafluorphenylacetic acid, benzeneacetic acid, 2,3,4,5,6-pentafluoro, acmc-20aoj2, pentafluorophenylaceticacid, 2-pentafluorophenyl acetic acid |
| PubChem CID | 69553 |
| IUPAC Name | 2-(2,3,4,5,6-pentafluorophenyl)acetic acid |
| SMILES | C(C1=C(C(=C(C(=C1F)F)F)F)F)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2243003
|
Thermo Scientific Chemicals
B2243003 |
1 g |
Each for $78.56
|
|
|||||
|
AAB2243006
|
Thermo Scientific Chemicals
B2243006 |
5 g |
Each for $250.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 653-21-4 | |
| 226.102 | |
| LGCODSNZJOVMHV-UHFFFAOYSA-N | |
| 69553 | |
| C(C1=C(C(=C(C(=C1F)F)F)F)F)C(=O)O |
| C8H3F5O2 | |
| MFCD00004316 | |
| pentafluorophenylacetic acid, 2,3,4,5,6-pentafluorophenylacetic acid, pentafluorophenyl acetic acid, 2-perfluorophenyl acetic acid, 2-2,3,4,5,6-pentafluorophenyl acetic acid, 2,3,4,5,6-pentafluorphenylacetic acid, benzeneacetic acid, 2,3,4,5,6-pentafluoro, acmc-20aoj2, pentafluorophenylaceticacid, 2-pentafluorophenyl acetic acid | |
| 2-(2,3,4,5,6-pentafluorophenyl)acetic acid |
Specifications
| 653-21-4 | |
| C8H3F5O2 | |
| 1 g | |
| pentafluorophenylacetic acid, 2,3,4,5,6-pentafluorophenylacetic acid, pentafluorophenyl acetic acid, 2-perfluorophenyl acetic acid, 2-2,3,4,5,6-pentafluorophenyl acetic acid, 2,3,4,5,6-pentafluorphenylacetic acid, benzeneacetic acid, 2,3,4,5,6-pentafluoro, acmc-20aoj2, pentafluorophenylaceticacid, 2-pentafluorophenyl acetic acid | |
| C(C1=C(C(=C(C(=C1F)F)F)F)F)C(=O)O | |
| 226.102 | |
| 226.1 | |
| 2,3,4,5,6-Pentafluorophenylacetic acid |
| 107°C to 110°C | |
| MFCD00004316 | |
| 2056612 | |
| LGCODSNZJOVMHV-UHFFFAOYSA-N | |
| 2-(2,3,4,5,6-pentafluorophenyl)acetic acid | |
| 69553 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-497-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only