missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorophenylacetic acid, 98+%

Catalog No. AAB2243003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2243003 1 g
AAB2243006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2243003 Supplier Thermo Scientific Chemicals Supplier No. B2243003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 653-21-4 | C8H3F5O2 | 226.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 653-21-4
Molecular Formula C8H3F5O2
Molecular Weight (g/mol) 226.102
MDL Number MFCD00004316
InChI Key LGCODSNZJOVMHV-UHFFFAOYSA-N
Synonym pentafluorophenylacetic acid, 2,3,4,5,6-pentafluorophenylacetic acid, pentafluorophenyl acetic acid, 2-perfluorophenyl acetic acid, 2-2,3,4,5,6-pentafluorophenyl acetic acid, 2,3,4,5,6-pentafluorphenylacetic acid, benzeneacetic acid, 2,3,4,5,6-pentafluoro, acmc-20aoj2, pentafluorophenylaceticacid, 2-pentafluorophenyl acetic acid
PubChem CID 69553
IUPAC Name 2-(2,3,4,5,6-pentafluorophenyl)acetic acid
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)C(=O)O

Specifications

Melting Point 107°C to 110°C
Quantity 1 g
Beilstein 2056612
Formula Weight 226.1
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5,6-Pentafluorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-497-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only