missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4,5,6-Pentafluorobenzyl Cyanide 97.0+%, TCI America™
$78.29 - $311.30
Chemical Identifiers
| CAS | 653-30-5 |
|---|---|
| Molecular Formula | C8H2F5N |
| Molecular Weight (g/mol) | 207.103 |
| MDL Number | MFCD00013814 |
| InChI Key | YDNOJUAQBFXZCR-UHFFFAOYSA-N |
| Synonym | 2,3,4,5,6-pentafluorophenylacetonitrile, pentafluorophenylacetonitrile, pentafluorophenyl acetonitrile, 2-2,3,4,5,6-pentafluorophenyl acetonitrile, 2,3,4,5,6-pentafluorobenzyl cyanide, 2-perfluorophenyl acetonitrile, benzeneacetonitrile, 2,3,4,5,6-pentafluoro, 2-2,3,4,5,6-pentafluorophenyl ethanenitrile, acmc-1ay9x, 2-pentafluorophenyl acetonitrile |
| PubChem CID | 69554 |
| IUPAC Name | 2-(2,3,4,5,6-pentafluorophenyl)acetonitrile |
| SMILES | C(C#N)C1=C(C(=C(C(=C1F)F)F)F)F |
Chemical Identifiers
| 653-30-5 | |
| 207.103 | |
| YDNOJUAQBFXZCR-UHFFFAOYSA-N | |
| 69554 | |
| C(C#N)C1=C(C(=C(C(=C1F)F)F)F)F |
| C8H2F5N | |
| MFCD00013814 | |
| 2,3,4,5,6-pentafluorophenylacetonitrile, pentafluorophenylacetonitrile, pentafluorophenyl acetonitrile, 2-2,3,4,5,6-pentafluorophenyl acetonitrile, 2,3,4,5,6-pentafluorobenzyl cyanide, 2-perfluorophenyl acetonitrile, benzeneacetonitrile, 2,3,4,5,6-pentafluoro, 2-2,3,4,5,6-pentafluorophenyl ethanenitrile, acmc-1ay9x, 2-pentafluorophenyl acetonitrile | |
| 2-(2,3,4,5,6-pentafluorophenyl)acetonitrile |
Specifications
| 653-30-5 | |
| 110°C | |
| MFCD00013814 | |
| 3276 | |
| YDNOJUAQBFXZCR-UHFFFAOYSA-N | |
| 2-(2,3,4,5,6-pentafluorophenyl)acetonitrile | |
| 69554 | |
| ≥97.0% (GC) | |
| 2,3,4,5,6-Pentafluorobenzyl Cyanide |
| Yellow | |
| C8H2F5N | |
| 1 g | |
| 2,3,4,5,6-pentafluorophenylacetonitrile, pentafluorophenylacetonitrile, pentafluorophenyl acetonitrile, 2-2,3,4,5,6-pentafluorophenyl acetonitrile, 2,3,4,5,6-pentafluorobenzyl cyanide, 2-perfluorophenyl acetonitrile, benzeneacetonitrile, 2,3,4,5,6-pentafluoro, 2-2,3,4,5,6-pentafluorophenyl ethanenitrile, acmc-1ay9x, 2-pentafluorophenyl acetonitrile | |
| C(C#N)C1=C(C(=C(C(=C1F)F)F)F)F | |
| 207.103 | |
| 207.10 | |
| Liquid |
Safety and Handling
TSCA : No