missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorobenzyl Cyanide 97.0+%, TCI America™

Catalog No. P22441G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22441G 1 g
P22445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22441G Supplier TCI America Supplier No. P22441G

Chemical Identifiers

CAS 653-30-5
Molecular Formula C8H2F5N
Molecular Weight (g/mol) 207.103
MDL Number MFCD00013814
InChI Key YDNOJUAQBFXZCR-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorophenylacetonitrile, pentafluorophenylacetonitrile, pentafluorophenyl acetonitrile, 2-2,3,4,5,6-pentafluorophenyl acetonitrile, 2,3,4,5,6-pentafluorobenzyl cyanide, 2-perfluorophenyl acetonitrile, benzeneacetonitrile, 2,3,4,5,6-pentafluoro, 2-2,3,4,5,6-pentafluorophenyl ethanenitrile, acmc-1ay9x, 2-pentafluorophenyl acetonitrile
PubChem CID 69554
IUPAC Name 2-(2,3,4,5,6-pentafluorophenyl)acetonitrile
SMILES C(C#N)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
UN Number 3276
Formula Weight 207.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl Cyanide