missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester, 96%
CAS: 325142-81-2 | C12H12BF5O2 | 294.028 g/mol
$190.66 - $628.45
Chemical Identifiers
| CAS | 325142-81-2 |
|---|---|
| Molecular Formula | C12H12BF5O2 |
| Molecular Weight (g/mol) | 294.028 |
| MDL Number | MFCD12405352 |
| InChI Key | FTJVUPKSBLYGSY-UHFFFAOYSA-N |
| Synonym | 4,4,5,5-tetramethyl-2-perfluorophenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-pentafluorophenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2,3,4,5,6-pentafluorophenyl-1,3,2-dioxaborolane, perfluorophenylboronic acid,pinacol ester, perfluorophenylbornic acid, pinacol ester, perfluorophenylboronic acid, pinacol ester, 2,3,4,5,6-pentafluorobenzeneboronic acid pinacol ester, 2,3,4,5,6-pentafluorophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2,3,4,5,6-pentafluorophenyl, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-2,3,4,5,6-pentafluorobenzene |
| PubChem CID | 23065713 |
| IUPAC Name | 4,4,5,5-tetramethyl-2-(2,3,4,5,6-pentafluorophenyl)-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C(=C2F)F)F)F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH60431MD
|
Thermo Scientific Chemicals
H60431MD |
250 mg |
Each for $190.66
|
|
|||||
|
AAH6043103
|
Thermo Scientific Chemicals
H6043103 |
1 g |
Each for $628.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 325142-81-2 | |
| 294.028 | |
| FTJVUPKSBLYGSY-UHFFFAOYSA-N | |
| 23065713 | |
| B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C(=C2F)F)F)F)F |
| C12H12BF5O2 | |
| MFCD12405352 | |
| 4,4,5,5-tetramethyl-2-perfluorophenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-pentafluorophenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2,3,4,5,6-pentafluorophenyl-1,3,2-dioxaborolane, perfluorophenylboronic acid,pinacol ester, perfluorophenylbornic acid, pinacol ester, perfluorophenylboronic acid, pinacol ester, 2,3,4,5,6-pentafluorobenzeneboronic acid pinacol ester, 2,3,4,5,6-pentafluorophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2,3,4,5,6-pentafluorophenyl, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-2,3,4,5,6-pentafluorobenzene | |
| 4,4,5,5-tetramethyl-2-(2,3,4,5,6-pentafluorophenyl)-1,3,2-dioxaborolane |
Specifications
| 325142-81-2 | |
| 66°C (0.4 mmHg) | |
| MFCD12405352 | |
| 4,4,5,5-tetramethyl-2-perfluorophenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-pentafluorophenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2,3,4,5,6-pentafluorophenyl-1,3,2-dioxaborolane, perfluorophenylboronic acid,pinacol ester, perfluorophenylbornic acid, pinacol ester, perfluorophenylboronic acid, pinacol ester, 2,3,4,5,6-pentafluorobenzeneboronic acid pinacol ester, 2,3,4,5,6-pentafluorophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2,3,4,5,6-pentafluorophenyl, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-2,3,4,5,6-pentafluorobenzene | |
| B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C(=C2F)F)F)F)F | |
| 294.028 | |
| 294.03 | |
| 2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester |
| 35°C to 36°C | |
| C12H12BF5O2 | |
| 250 mg | |
| FTJVUPKSBLYGSY-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-(2,3,4,5,6-pentafluorophenyl)-1,3,2-dioxaborolane | |
| 23065713 | |
| 96% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only