Learn More
2,3,4,5,6-Pentafluoroanisole, 98%
CAS: 389-40-2 | C7H3F5O | 198.092 g/mol
Supplier: Thermo Scientific Chemicals B2392814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,3,4,5,6-Pentafluoroanisole | |
| -38°C | |
| 136°C to 138°C | |
| C7H3F5O | |
| MFCD00000289 | |
| UN1993 | |
| 2,3,4,5,6-pentafluoroanisole, pentafluoroanisole, benzene, pentafluoromethoxy, pentafluorophenyl methyl ether, pentafluoromethoxybenzene, methyl pentafluorophenyl ether, benzene, 1,2,3,4,5-pentafluoro-6-methoxy, anisole, 2,3,4,5,6-pentafluoro, 1,2,3,4,6-pentafluoro-5-methoxybenzene, methoxypentafluorobenzene | |
| COC1=C(C(=C(C(=C1F)F)F)F)F | |
| 198.092 | |
| 198.09 |
| 389-40-2 | |
| 1.5 | |
| 32°C (89°F) | |
| 1.409 | |
| 25 g | |
| 2052664 | |
| ZRQUIRABLIQJRI-UHFFFAOYSA-N | |
| 1,2,3,4,5-pentafluoro-6-methoxybenzene | |
| 67849 | |
| 98% |
Chemical Identifiers
| 389-40-2 | |
| 198.092 | |
| ZRQUIRABLIQJRI-UHFFFAOYSA-N | |
| 67849 | |
| COC1=C(C(=C(C(=C1F)F)F)F)F |
| C7H3F5O | |
| MFCD00000289 | |
| 2,3,4,5,6-pentafluoroanisole, pentafluoroanisole, benzene, pentafluoromethoxy, pentafluorophenyl methyl ether, pentafluoromethoxybenzene, methyl pentafluorophenyl ether, benzene, 1,2,3,4,5-pentafluoro-6-methoxy, anisole, 2,3,4,5,6-pentafluoro, 1,2,3,4,6-pentafluoro-5-methoxybenzene, methoxypentafluorobenzene | |
| 1,2,3,4,5-pentafluoro-6-methoxybenzene |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 206-866-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only