missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',3',4',5',6'-Pentafluoroacetophenone 98.0+%, TCI America™
Supplier: TCI America P09215G
Specifications
| 2′,3′,4′,5′,6′-Pentafluoroacetophenone | |
| Yellow | |
| C8H3F5O | |
| 5 g | |
| FBGHCYZBCMDEOX-UHFFFAOYSA-N | |
| 1-(2,3,4,5,6-pentafluorophenyl)ethan-1-one | |
| 69546 | |
| ≥98.0% (GC) |
| 652-29-9 | |
| 131°C | |
| MFCD00000296 | |
| 2',3',4',5',6'-pentafluoroacetophenone, 1-perfluorophenyl ethanone, pentafluoroacetophenone, 1-pentafluorophenyl ethan-1-one, ethanone, 1-pentafluorophenyl, 2,3,4,5,6-pentafluoroacetophenone, 1-pentafluorphenyl ethanon, 1-acetyl-2,3,4,5,6-pentafluorobenzene, acmc-209npz, 1-pentafluorophenylethanone | |
| CC(=O)C1=C(F)C(F)=C(F)C(F)=C1F | |
| 210.10 | |
| 210.10 | |
| Liquid |
Chemical Identifiers
| 652-29-9 | |
| 210.10 | |
| FBGHCYZBCMDEOX-UHFFFAOYSA-N | |
| 69546 | |
| CC(=O)C1=C(F)C(F)=C(F)C(F)=C1F |
| C8H3F5O | |
| MFCD00000296 | |
| 2',3',4',5',6'-pentafluoroacetophenone, 1-perfluorophenyl ethanone, pentafluoroacetophenone, 1-pentafluorophenyl ethan-1-one, ethanone, 1-pentafluorophenyl, 2,3,4,5,6-pentafluoroacetophenone, 1-pentafluorphenyl ethanon, 1-acetyl-2,3,4,5,6-pentafluorobenzene, acmc-209npz, 1-pentafluorophenylethanone | |
| 1-(2,3,4,5,6-pentafluorophenyl)ethan-1-one |
Safety and Handling
TSCA : No