accessibility menu, dialog, popup

UserName

H3COFFFFF

CAS RN 652-29-9

IUPAC Name: 1-(2,3,4,5,6-pentafluorophenyl)ethan-1-one
Synonyms: pentafluoroacetophenone
Molecular Weight (g/mol): 210.1
Molecular Formula: C8H3F5O
InChi Key: FBGHCYZBCMDEOX-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C(F)=C(F)C(F)=C1F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)