missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',3',4',5',6'-Pentafluoroacetophenone, 97%
CAS: 652-29-9 | C8H3F5O | 210.10 g/mol
$65.32 - $65.32
Chemical Identifiers
| CAS | 652-29-9 |
|---|---|
| Molecular Formula | C8H3F5O |
| Molecular Weight (g/mol) | 210.10 |
| MDL Number | MFCD00000296 |
| InChI Key | FBGHCYZBCMDEOX-UHFFFAOYSA-N |
| Synonym | 2',3',4',5',6'-pentafluoroacetophenone, 1-perfluorophenyl ethanone, pentafluoroacetophenone, 1-pentafluorophenyl ethan-1-one, ethanone, 1-pentafluorophenyl, 2,3,4,5,6-pentafluoroacetophenone, 1-pentafluorphenyl ethanon, 1-acetyl-2,3,4,5,6-pentafluorobenzene, acmc-209npz, 1-pentafluorophenylethanone |
| PubChem CID | 69546 |
| SMILES | CC(=O)C1=C(F)C(F)=C(F)C(F)=C1F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0206303
|
Thermo Scientific Chemicals
L0206303 |
1 g |
Each for $65.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 652-29-9 | |
| 210.10 | |
| FBGHCYZBCMDEOX-UHFFFAOYSA-N | |
| 69546 |
| C8H3F5O | |
| MFCD00000296 | |
| 2',3',4',5',6'-pentafluoroacetophenone, 1-perfluorophenyl ethanone, pentafluoroacetophenone, 1-pentafluorophenyl ethan-1-one, ethanone, 1-pentafluorophenyl, 2,3,4,5,6-pentafluoroacetophenone, 1-pentafluorphenyl ethanon, 1-acetyl-2,3,4,5,6-pentafluorobenzene, acmc-209npz, 1-pentafluorophenylethanone | |
| CC(=O)C1=C(F)C(F)=C(F)C(F)=C1F |
Specifications
| 652-29-9 | |
| 130°C to 131°C | |
| C8H3F5O | |
| MFCD00000296 | |
| 2053225 | |
| FBGHCYZBCMDEOX-UHFFFAOYSA-N | |
| 1-(2,3,4,5,6-pentafluorophenyl)ethanone | |
| 69546 | |
| 97% |
| 1.476 | |
| 65°C (149°F) | |
| 1.4366 | |
| 1 g | |
| 2',3',4',5',6'-pentafluoroacetophenone, 1-perfluorophenyl ethanone, pentafluoroacetophenone, 1-pentafluorophenyl ethan-1-one, ethanone, 1-pentafluorophenyl, 2,3,4,5,6-pentafluoroacetophenone, 1-pentafluorphenyl ethanon, 1-acetyl-2,3,4,5,6-pentafluorobenzene, acmc-209npz, 1-pentafluorophenylethanone | |
| CC(=O)C1=C(F)C(F)=C(F)C(F)=C1F | |
| 210.10 | |
| 210.1 | |
| 2',3',4',5',6'-Pentafluoroacetophenone |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 211-487-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only