missing translation for 'onlineSavingsMsg'
Learn More

2,3,3-Trimethyl-3H-benzo[g]indole 98.0+%, TCI America™

Supplier:  TCI America T260125G

 View more versions of this product

Catalog No. T260125G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3,3-Trimethyl-3H-benzo[g]indole
78°C
C15H15N
25 g
MNRRNPKQXGBGBH-UHFFFAOYSA-N
2,3,3-trimethylbenzo[g]indole
12871551
≥98.0% (HPLC,T)
74470-85-2
127°C
MFCD00723378
2,3,3-trimethyl-3h-benzo g indole, 2,3,3-trimethylbenzo g indole, 2,3,3-trimethyl-3h-benz g indole, 3h-benz g indole, 2,3,3-trimethyl, 2,3,3-trimethyl-3h-benz g indol, acmc-209ouz, 2,3,3-trimethylbenzindole, 2,3,3-trimethyl-6,7-benzoindolenine, 2,3,3-trimethyl-3-hydrobenzo g indole
CC1=NC2=C(C1(C)C)C=CC3=CC=CC=C32
209.292
209.29
Crystalline Powder

Chemical Identifiers

74470-85-2
209.292
MNRRNPKQXGBGBH-UHFFFAOYSA-N
12871551
CC1=NC2=C(C1(C)C)C=CC3=CC=CC=C32
C15H15N
MFCD00723378
2,3,3-trimethyl-3h-benzo g indole, 2,3,3-trimethylbenzo g indole, 2,3,3-trimethyl-3h-benz g indole, 3h-benz g indole, 2,3,3-trimethyl, 2,3,3-trimethyl-3h-benz g indol, acmc-209ouz, 2,3,3-trimethylbenzindole, 2,3,3-trimethyl-6,7-benzoindolenine, 2,3,3-trimethyl-3-hydrobenzo g indole
2,3,3-trimethylbenzo[g]indole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions