accessibility menu, dialog, popup

UserName

CAS RN 74470-85-2

IUPAC Name: 2,3,3-trimethyl-3H-benzo[g]indole
Synonyms: 2,3,3-trimethylbenzo[g]indole
Molecular Weight (g/mol): 209.29
Molecular Formula: C15H15N
InChi Key: MNRRNPKQXGBGBH-UHFFFAOYSA-N
SMILES: CC1=NC2=C3C=CC=CC3=CC=C2C1(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results