missing translation for 'onlineSavingsMsg'
Learn More

2,3,3-Trimethyl-3H-benzo[g]indole 98.0+%, TCI America™

Catalog No. T26015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26015G 5 g
T260125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26015G Supplier TCI America Supplier No. T26015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74470-85-2
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.292
MDL Number MFCD00723378
InChI Key MNRRNPKQXGBGBH-UHFFFAOYSA-N
Synonym 2,3,3-trimethyl-3h-benzo g indole, 2,3,3-trimethylbenzo g indole, 2,3,3-trimethyl-3h-benz g indole, 3h-benz g indole, 2,3,3-trimethyl, 2,3,3-trimethyl-3h-benz g indol, acmc-209ouz, 2,3,3-trimethylbenzindole, 2,3,3-trimethyl-6,7-benzoindolenine, 2,3,3-trimethyl-3-hydrobenzo g indole
PubChem CID 12871551
IUPAC Name 2,3,3-trimethylbenzo[g]indole
SMILES CC1=NC2=C(C1(C)C)C=CC3=CC=CC=C32

Specifications

Melting Point 78°C
Boiling Point 127°C
Quantity 5 g
Formula Weight 209.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,3-Trimethyl-3H-benzo[g]indole