missing translation for 'onlineSavingsMsg'
Learn More

2,3,3,5-Tetramethylindolenine 98.0+%, TCI America™

Catalog No. p-7137762
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
T34585G 5 g
T345825G 25 g
2 options

Catalog No. T34585G

Supplier: TCI America T34585G

Only null left

Chemical Identifiers

CAS 25981-82-2
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.26
MDL Number MFCD03093020
InChI Key RQVAPBRSUHSDGP-UHFFFAOYSA-N
PubChem CID 117693
IUPAC Name 2,3,3,5-tetramethyl-3H-indole
SMILES CC1=NC2=CC=C(C)C=C2C1(C)C

Specifications

Boiling Point 125°C
Quantity 5 g
Synonym 2,3,3,5-tetramethyl-3h-indole, 2,3,3,5-tetramethylindolenine, 3h-indole, 2,3,3,5-tetramethyl, pubchem2092, 2,3,3,5-tetramethyl-3h-indol, 2,3,3,5-tetramethyl-3-h-indole, 3h-indole,2,3,3,5-tetramethyl, 2,3,3,5-tetramethyl-3-hydroindole, 2,3,3,5-tetramethyl-3-h indole
Formula Weight 173.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,3,5-Tetramethylindolenine