accessibility menu, dialog, popup

UserName

CH3NH3CH3CCH3

CAS RN 25981-82-2

IUPAC Name: 2,3,3,5-tetramethyl-3H-indole
Synonyms: 2,3,3,5-tetramethylindole
Molecular Weight (g/mol): 173.26
Molecular Formula: C12H15N
InChi Key: RQVAPBRSUHSDGP-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C(C)C=C2C1(C)C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)