missing translation for 'onlineSavingsMsg'
Learn More

2,3,3,5-Tetramethylindolenine 98.0+%, TCI America™

Catalog No. T34585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34585G 5 g
T345825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34585G Supplier TCI America Supplier No. T34585G

Chemical Identifiers

CAS 25981-82-2
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.26
MDL Number MFCD03093020
InChI Key RQVAPBRSUHSDGP-UHFFFAOYSA-N
PubChem CID 117693
IUPAC Name 2,3,3,5-tetramethyl-3H-indole
SMILES CC1=NC2=CC=C(C)C=C2C1(C)C

Specifications

Boiling Point 125°C
Quantity 5 g
Synonym 2,3,3,5-tetramethyl-3h-indole, 2,3,3,5-tetramethylindolenine, 3h-indole, 2,3,3,5-tetramethyl, pubchem2092, 2,3,3,5-tetramethyl-3h-indol, 2,3,3,5-tetramethyl-3-h-indole, 3h-indole,2,3,3,5-tetramethyl, 2,3,3,5-tetramethyl-3-hydroindole, 2,3,3,5-tetramethyl-3-h indole
Formula Weight 173.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,3,5-Tetramethylindolenine