missing translation for 'onlineSavingsMsg'
Learn More

2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J64981LB0

 View more versions of this product

Catalog No. AAJ64981LB0

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride
White
MFCD06410993
2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, c11h15cl2no2.hcl, 2-3-2,3-dichlorophenoxy propylamino ethanol hydrochloride
QVEIRCZEBQRCTR-UHFFFAOYSA-O
2-[3-(2,3-dichlorophenoxy)propylamino]ethanol;hydrochloride
52913859
Powder
418788-90-6
C11H16Cl2NO2
5 mg
Soluble in water at ∽25mg/ml; Also soluble in ethanol,DMSO or DMF at ∽20mg/ml; Soluble in PBS,pH 7;2 at ∽10mg/ml.
OCC[NH2+]CCCOC1=CC=CC(Cl)=C1Cl
265.15
300.61

Chemical Identifiers

418788-90-6
265.15
QVEIRCZEBQRCTR-UHFFFAOYSA-O
52913859
OCC[NH2+]CCCOC1=CC=CC(Cl)=C1Cl
C11H16Cl2NO2
MFCD06410993
2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, c11h15cl2no2.hcl, 2-3-2,3-dichlorophenoxy propylamino ethanol hydrochloride
2-[3-(2,3-dichlorophenoxy)propylamino]ethanol;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only