CAS RN 418788-90-6
CAS RN 418788-90-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride, Thermo Scientific Chemicals
CAS: 418788-90-6 Molecular Formula: C11H16Cl2NO2 Molecular Weight (g/mol): 265.15 MDL Number: MFCD06410993 InChI Key: QVEIRCZEBQRCTR-UHFFFAOYSA-O Synonym: 2,3-dcpe hydrochloride,2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride,c11h15cl2no2.hcl,2-3-2,3-dichlorophenoxy propylamino ethanol hydrochloride PubChem CID: 52913859 IUPAC Name: 2-[3-(2,3-dichlorophenoxy)propylamino]ethanol;hydrochloride SMILES: OCC[NH2+]CCCOC1=CC=CC(Cl)=C1Cl