missing translation for 'onlineSavingsMsg'
Learn More

2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride, Thermo Scientific Chemicals

Catalog No. AAJ64981MA
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64981MA 10 mg
AAJ64981LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64981MA Supplier Thermo Scientific Chemicals Supplier No. J64981MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 418788-90-6 | Molecular Formula: C11H16Cl2NO2 | Molecular Weight: 265.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 418788-90-6
Molecular Formula C11H16Cl2NO2
Molecular Weight (g/mol) 265.15
MDL Number MFCD06410993
InChI Key QVEIRCZEBQRCTR-UHFFFAOYSA-O
Synonym 2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, c11h15cl2no2.hcl, 2-3-2,3-dichlorophenoxy propylamino ethanol hydrochloride
PubChem CID 52913859
IUPAC Name 2-[3-(2,3-dichlorophenoxy)propylamino]ethanol;hydrochloride
SMILES OCC[NH2+]CCCOC1=CC=CC(Cl)=C1Cl

Specifications

Color White
Quantity 10 mg
Solubility Information Soluble in water at ∽25mg/ml; Also soluble in ethanol,DMSO or DMF at ∽20mg/ml; Soluble in PBS,pH 7;2 at ∽10mg/ml.
Formula Weight 300.61
Physical Form Powder
Chemical Name or Material 2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only