missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Thiobis(4-tert-octylphenol) 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 137°C |
| Color | White-Yellow |
| Quantity | 500 g |
| Formula Weight | 442.70 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2′-Thiobis(4-tert-octylphenol) |
Safety and Handling
| EINECSNumber | (3)-1113 |
| TSCA | Yes |