missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Thiobis(4-tert-octylphenol) 95.0+%, TCI America™

Catalog No. T167425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T167425G 25 g
T1674500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T167425G Supplier TCI America Supplier No. T167425G
Only null left

Chemical Identifiers

CAS 3294-03-9
Molecular Formula C28H42O2S
Molecular Weight (g/mol) 442.70
MDL Number MFCD00190577
InChI Key WQYFETFRIRDUPJ-UHFFFAOYSA-N
Synonym Bis(2-hydroxy-5-tert-octylphenyl) Sulfide
PubChem CID 76798
IUPAC Name 2-[2-hydroxy-5-(2,4,4-trimethylpentan-2-yl)phenyl]sulfanyl-4-(2,4,4-trimethylpentan-2-yl)phenol
SMILES CC(C)(C)CC(C)(C)C1=CC(=C(C=C1)O)SC2=C(C=CC(=C2)C(C)(C)CC(C)(C)C)O

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 25 g
Formula Weight 442.70
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Thiobis(4-tert-octylphenol)
EINECSNumber (3)-1113
TSCA Yes