missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienyl)pyridine, 97%

CAS: 3319-99-1 | C9H7NS | 161.22 g/mol

Supplier:  Thermo Scientific Chemicals L0477506

 View more versions of this product

Catalog No. AAL0477506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-(2-Thienyl)pyridine
60°C to 65°C
MFCD00044441
508454
QLPKTAFPRRIFQX-UHFFFAOYSA-N
2-thiophen-2-ylpyridine
76832
97%
3319-99-1
C9H7NS
5 g
2-2-thienyl pyridine, 2-thiophen-2-yl pyridine, 2-thien-2-ylpyridine, 2-2-pyridyl thiophene, pyridine, 2-2-thienyl, thienyl pyridine, 2-thienylpyridine, 2-pyrid-2-ylthiophene, acmc-1cknd, 2-thien-2-yl pyridine
S1C=CC=C1C1=CC=CC=N1
161.22
161.23

Chemical Identifiers

3319-99-1
161.22
QLPKTAFPRRIFQX-UHFFFAOYSA-N
76832
S1C=CC=C1C1=CC=CC=N1
C9H7NS
MFCD00044441
2-2-thienyl pyridine, 2-thiophen-2-yl pyridine, 2-thien-2-ylpyridine, 2-2-pyridyl thiophene, pyridine, 2-2-thienyl, thienyl pyridine, 2-thienylpyridine, 2-pyrid-2-ylthiophene, acmc-1cknd, 2-thien-2-yl pyridine
2-thiophen-2-ylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 222-022-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only