missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienyl)pyridine, 97%

Catalog No. p-7044135
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0477503 1 g
AAL0477506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0477503 Supplier Thermo Scientific Chemicals Supplier No. L0477503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3319-99-1 | C9H7NS | 161.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3319-99-1
Molecular Formula C9H7NS
Molecular Weight (g/mol) 161.22
MDL Number MFCD00044441
InChI Key QLPKTAFPRRIFQX-UHFFFAOYSA-N
Synonym 2-2-thienyl pyridine, 2-thiophen-2-yl pyridine, 2-thien-2-ylpyridine, 2-2-pyridyl thiophene, pyridine, 2-2-thienyl, thienyl pyridine, 2-thienylpyridine, 2-pyrid-2-ylthiophene, acmc-1cknd, 2-thien-2-yl pyridine
PubChem CID 76832
IUPAC Name 2-thiophen-2-ylpyridine
SMILES S1C=CC=C1C1=CC=CC=N1

Specifications

Melting Point 60°C to 65°C
Quantity 1 g
Beilstein 508454
Formula Weight 161.23
Percent Purity 97%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2-(2-Thienyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-022-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only