missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Thienyl)-1,3-dioxolane 97.0+%, TCI America™
Supplier: TCI America T20495G
Specifications
| 2-(2-Thienyl)-1,3-dioxolane | |
| Yellow | |
| C7H8O2S | |
| 5 g | |
| FHPFDCRNWLVVHD-UHFFFAOYSA-N | |
| 2-thiophen-2-yl-1,3-dioxolane | |
| 3653561 | |
| ≥97.0% (GC) |
| 58268-08-9 | |
| 111°C | |
| MFCD01924576 | |
| 2-(1,3-Dioxolan-2-yl)thiophene | |
| C1COC(O1)C2=CC=CS2 | |
| 156.199 | |
| 156.20 | |
| Liquid |
Chemical Identifiers
| 58268-08-9 | |
| 156.199 | |
| FHPFDCRNWLVVHD-UHFFFAOYSA-N | |
| 3653561 | |
| C1COC(O1)C2=CC=CS2 |
| C7H8O2S | |
| MFCD01924576 | |
| 2-(1,3-Dioxolan-2-yl)thiophene | |
| 2-thiophen-2-yl-1,3-dioxolane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator