missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Thienyl)-1,3-dioxolane 97.0+%, TCI America™
$109.98 - $324.35
Chemical Identifiers
| CAS | 58268-08-9 |
|---|---|
| Molecular Formula | C7H8O2S |
| Molecular Weight (g/mol) | 156.199 |
| MDL Number | MFCD01924576 |
| InChI Key | FHPFDCRNWLVVHD-UHFFFAOYSA-N |
| Synonym | 2-(1,3-Dioxolan-2-yl)thiophene |
| PubChem CID | 3653561 |
| IUPAC Name | 2-thiophen-2-yl-1,3-dioxolane |
| SMILES | C1COC(O1)C2=CC=CS2 |
Chemical Identifiers
| 58268-08-9 | |
| 156.199 | |
| FHPFDCRNWLVVHD-UHFFFAOYSA-N | |
| 3653561 | |
| C1COC(O1)C2=CC=CS2 |
| C7H8O2S | |
| MFCD01924576 | |
| 2-(1,3-Dioxolan-2-yl)thiophene | |
| 2-thiophen-2-yl-1,3-dioxolane |
Specifications
| 58268-08-9 | |
| 111°C | |
| MFCD01924576 | |
| 2-(1,3-Dioxolan-2-yl)thiophene | |
| C1COC(O1)C2=CC=CS2 | |
| 156.199 | |
| 156.20 | |
| Liquid |
| Yellow | |
| C7H8O2S | |
| 5 g | |
| FHPFDCRNWLVVHD-UHFFFAOYSA-N | |
| 2-thiophen-2-yl-1,3-dioxolane | |
| 3653561 | |
| ≥97.0% (GC) | |
| 2-(2-Thienyl)-1,3-dioxolane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator