missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Pyrrolidinyl)pyridine, 96%

Catalog No. p-7033149
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50176MD 250 mg
AAH5017603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50176MD Supplier Thermo Scientific Chemicals Supplier No. H50176MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77790-61-5 | C9H12N2 | 148.209 g/mol

2-(2-Pyrrolidinyl)pyridine is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Pyrrolidinyl)pyridine is used as primary and secondary intermediate.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 77790-61-5
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD01691538
InChI Key NDCZQFDBSPOUDF-UHFFFAOYSA-N
Synonym 2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine
PubChem CID 2771659
IUPAC Name 2-pyrrolidin-2-ylpyridine
SMILES C1CC(NC1)C2=CC=CC=N2

Specifications

Quantity 250 mg
UN Number UN2810
Solubility Information Soluble in water.
Formula Weight 148.21
Percent Purity 96%
Chemical Name or Material 2-(2-Pyrrolidinyl)pyridine
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECSNumber UT7650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only