missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Pyridyl)benzimidazole, 99%

Catalog No. AC167950100
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167950100 10 g Glass bottle
AC167950050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167950100 Supplier Thermo Scientific Chemicals Supplier No. 167950100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1137-68-4 | C12H9N3 | 195.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1137-68-4
Molecular Formula C12H9N3
Molecular Weight (g/mol) 195.23
MDL Number MFCD00005586
InChI Key YNFBMDWHEHETJW-UHFFFAOYSA-N
Synonym 2-2-pyridyl benzimidazole, 2-pyridin-2-yl-1h-benzo d imidazole, 2-pyridin-2-yl-1h-benzoimidazole, 2-pyridin-2-yl-1h-1,3-benzodiazole, 2-pyridin-2-yl-1h-benzimidazole, chembl72683, 1h-benzimidazole, 2-pyridinyl, 1h-benzimidazole, 2-2-pyridinyl, acmc-20aime, 2 2-pyridyl benzimidazole
PubChem CID 70821
IUPAC Name 2-pyridin-2-yl-1H-benzimidazole
SMILES N1C2=CC=CC=C2N=C1C1=CC=CC=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 220.0°C to 225.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: insoluble.
Formula Weight 195.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-(2-Pyridyl)benzimidazole, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 214-508-7
TSCA TSCA

RUO – Research Use Only