accessibility menu, dialog, popup

UserName

HNNN

CAS RN 1137-68-4

IUPAC Name: 2-(pyridin-2-yl)-1H-1,3-benzodiazole
Synonyms: 2-(pyridin-2-yl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 195.23
Molecular Formula: C12H9N3
InChi Key: YNFBMDWHEHETJW-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2N=C1C1=CC=CC=N1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)