missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Pyridyl)benzimidazole, 98+%
Description
2-(2-Pyridyl)benzimidazole is used as a chelating fluorescent ligand in the synthesis of cobalt(II) complex. It was also used in the preparation of (H(2)L(1))(2)[Ru(III)Cl(4)(CH(3)CN)(2)](2)[Ru(IV)Cl(4)(CH(3)CN)(2)]·2Cl·6H(2)O complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(2-Pyridyl)benzimidazole is used as a chelating fluorescent ligand in the synthesis of cobalt(II) complex. It was also used in the preparation of (H(2)L(1))(2)[Ru(III)Cl(4)(CH(3)CN)(2)](2)[Ru(IV)Cl(4)(CH(3)CN)(2)]·2Cl·6H(2)O complex.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-(2-Pyridyl)benzimidazole is used as a chelating fluorescent ligand in the synthesis of cobalt(II) complex. It was also used in the preparation of (H(2)L(1))(2)[Ru(III)Cl(4)(CH(3)CN)(2)](2)[Ru(IV)Cl(4)(CH(3)CN)(2)]·2Cl·6H(2)O complex.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 221°C to 225°C |
| Quantity | 1 g |
| Beilstein | 151411 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 195.22 |
| Percent Purity | ≥98% |
| Physical Form | Crystals or Powder or Crystalline Powder |
| Chemical Name or Material | 2-(2-Pyridyl)benzimidazole |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 214-508-7 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only