missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Pyridyl)benzimidazole 98.0+%, TCI America™

Catalog No. P15991G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P15991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P15991G Supplier TCI America Supplier No. P15991G

Chemical Identifiers

CAS 1137-68-4
Molecular Formula C12H9N3
Molecular Weight (g/mol) 195.23
MDL Number MFCD00005586
InChI Key YNFBMDWHEHETJW-UHFFFAOYSA-N
Synonym 2-2-pyridyl benzimidazole, 2-pyridin-2-yl-1h-benzo d imidazole, 2-pyridin-2-yl-1h-benzoimidazole, 2-pyridin-2-yl-1h-1,3-benzodiazole, 2-pyridin-2-yl-1h-benzimidazole, chembl72683, 1h-benzimidazole, 2-pyridinyl, 1h-benzimidazole, 2-2-pyridinyl, acmc-20aime, 2 2-pyridyl benzimidazole
PubChem CID 70821
IUPAC Name 2-(pyridin-2-yl)-1H-1,3-benzodiazole
SMILES N1C2=CC=CC=C2N=C1C1=CC=CC=N1

Specifications

Melting Point 225°C
Quantity 1 g
Formula Weight 195.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Pyridyl)benzimidazole