missing translation for 'onlineSavingsMsg'
Learn More

2,2-Pentamethylene-1,3-dioxolan-4-one 98.0+%, TCI America™

Catalog No. P10195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10195G 5 g
P101925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10195G Supplier TCI America Supplier No. P10195G
Only null left

Chemical Identifiers

CAS 4423-79-4
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.18
MDL Number MFCD00010852
InChI Key HBGAMCTZOLJGAS-UHFFFAOYSA-N
Synonym 1,4-Dioxaspiro[4.5]-2-decanone
PubChem CID 544223
IUPAC Name 1,4-dioxaspiro[4.5]decan-2-one
SMILES O=C1COC2(CCCCC2)O1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 156.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,2-Pentamethylene-1,3-dioxolan-4-one