accessibility menu, dialog, popup

UserName

OOO

CAS RN 4423-79-4

IUPAC Name: 1,4-dioxaspiro[4.5]decan-2-one
Synonyms: 1,4-dioxaspiro[4.5]decan-2-one
Molecular Weight (g/mol): 156.18
Molecular Formula: C8H12O3
InChi Key: HBGAMCTZOLJGAS-UHFFFAOYSA-N
SMILES: O=C1COC2(CCCCC2)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results