missing translation for 'onlineSavingsMsg'
Learn More

2,2'-O-Anhydro-5-methyluridine 98.0+%, TCI America™

Catalog No. A24311G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24311G Supplier TCI America Supplier No. A24311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22423-26-3
Molecular Formula C10H12N2O5
Molecular Weight (g/mol) 240.215
MDL Number MFCD00233555
InChI Key WLLOAUCNUMYOQI-JAGXHNFQSA-N
Synonym 2,2′C-O-Anhydro-(1-beta-D-arabinofuranosyl)-5-methyluracil
PubChem CID 168045
IUPAC Name (2R,3R,3aS,9aR)-3-hydroxy-2-(hydroxymethyl)-7-methyl-2,3,3a,9a-tetrahydrofuro[1,2][1,3]oxazolo[3,4-a]pyrimidin-6-one
SMILES CC1=CN2C3C(C(C(O3)CO)O)OC2=NC1=O

Specifications

Melting Point 219°C
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 240.22
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material 2,2′-O-Anhydro-5-methyluridine